Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papola | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011112.3 | NP_035242.1 | 22 | 2377 | ||
GenBank mRNA | AK090096.1 | - | 22 | 2378 | ||
AK132704.1 | - | 22 | 2364 | |||
BC060128.1 | - | 21 | 2224 |