Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plau | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008873.3 | NP_032899.1 | 8 | 839 | ||
GenBank mRNA | BC120709.1 | - | 7 | 779 | 40129632 | |
BC120713.1 | - | 7 | 779 | 40129636 | ||
X02389.1 | - | 8 | 794 |