Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011116.1 | NP_035246.1 | 11 | 1367 | ||
GenBank mRNA | AF026124.1 | - | 11 | 1367 | ||
AK153508.1 | - | 9 | 1056 | |||
AK188215.1 | - | 1 | 18 | |||
BC068170.1 | - | 11 | 1363 | |||
BC076586.1 | - | 11 | 1431 | 30604186 |