Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011122.3 | NP_035252.1 | 11, 12 | 1311 | ||
GenBank mRNA | AF046782.1 | - | 11, 12 | 1239 | ||
AK137480.1 | - | 11, 12 | 1262 | |||
BC006599.1 | - | 11, 12 | 1236 | 3965301 | ||
BC010268.1 | - | 3, 4 | 256 | 3589182 |