Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008886.2 | NP_032912.2 | 11 | 1758 | ||
GenBank mRNA | AK034866.1 | - | 11 | 1734 | ||
AK083049.1 | - | 11 | 1715 | |||
BC022179.1 | - | 11 | 1726 | |||
BC058189.1 | - | 11 | 1766 | |||
BC066110.1 | - | 11 | 1758 | |||
BC083502.1 | - | 11 | 1718 | |||
BC086637.1 | - | 11 | 1734 | |||
BC110684.1 | - | 11 | 1711 | |||
BC141287.1 | - | 11 | 1734 | 9055899 |