Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008886.2 | NP_032912.2 | 12 | 2161 | ||
GenBank mRNA | AK083049.1 | - | 12 | 2118 | ||
AK146365.1 | - | 12 | 2174 | |||
BC058189.1 | - | 12 | 2169 | |||
BC066110.1 | - | 12 | 2161 | |||
BC083502.1 | - | 12 | 2121 | |||
BC086637.1 | - | 12 | 2137 | |||
BC110684.1 | - | 12 | 2114 | |||
BC139774.1 | - | 12 | 1595 | |||
BC141287.1 | - | 12 | 2137 | 9055899 |