Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnliprp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018874.2 | NP_061362.1 | 12 | 1396 | ||
GenBank mRNA | AF061274.1 | - | 12 | 1397 | ||
AK028105.1 | - | 12 | 1309 | |||
BC068266.1 | - | 8 | 952 | |||
BC090985.1 | - | 12 | 1308 | 30315884 |