Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pold1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011131.3 | NP_035261.3 | 26 | 3255 | ||
GenBank mRNA | AK088310.1 | - | 26 | 3246 | ||
AK146837.1 | - | 26 | 3252 | |||
AK166592.1 | - | 25 | 3335 | |||
AK167569.1 | - | 26 | 3238 | |||
AK168967.1 | - | 26 | 3218 | |||
AK169040.1 | - | 26 | 3251 | |||
BC009128.1 | - | 26 | 3256 | 3257174 | ||
BC052670.1 | - | 25 | 3222 | 30058314 | ||
BX634546.1 | - | 2 | 170 | |||
Z21848.1 | - | 26 | 3224 |