Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppfibp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163557.1 | NP_001157029.1 | 23 | 2391 | ||
NM_008905.2 | NP_032931.2 | 24 | 2660 | |||
GenBank mRNA | AK012152.1 | - | 20 | 2049 | ||
AK166921.1 | - | 14 | 1528 | |||
AK185190.1 | - | 3 | 217 | |||
BC003966.1 | - | 9 | 957 | |||
BC055935.1 | - | 13 | 1261 | |||
BC100346.1 | - | 23 | 2391 | 30936010 | ||
U79024.1 | - | 24 | 2445 |