Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008909.2 | NP_032935.2 | 3 | 361 | ||
GenBank mRNA | AF126834.1 | - | 3 | 301 | ||
AK142562.1 | - | 3 | 356 | |||
BC057152.1 | - | 3 | 290 |