Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011156.2 | NP_035286.1 | 3 | 290 | ||
GenBank mRNA | AB007631.1 | - | 3 | 173 | ||
AK053816.1 | - | 3 | 301 | |||
AK076257.1 | - | 3 | 289 | |||
AK133875.1 | - | 3 | 288 | |||
AK141851.1 | - | 3 | 299 | |||
AK161528.1 | - | 3 | 289 | |||
AK163787.1 | - | 3 | 302 | |||
AK170570.1 | - | 3 | 299 | |||
BC012869.1 | - | 3 | 290 | 3967219 | ||
BC050830.1 | - | 3 | 283 | 6519113 |