Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011156.2 | NP_035286.1 | 7 | 851 | ||
GenBank mRNA | AB007631.1 | - | 7 | 734 | ||
AK053816.1 | - | 7 | 862 | |||
AK076257.1 | - | 7 | 850 | |||
AK133875.1 | - | 7 | 849 | |||
AK141851.1 | - | 7 | 860 | |||
AK161528.1 | - | 7 | 850 | |||
AK163787.1 | - | 7 | 863 | |||
AK170570.1 | - | 7 | 860 | |||
BC012869.1 | - | 7 | 851 | 3967219 | ||
BC050830.1 | - | 7 | 844 | 6519113 |