Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008922.2 | NP_032948.1 | 10 | 1161 | ||
GenBank mRNA | AK076095.1 | - | 10 | 1143 | ||
AK077760.1 | - | 10 | 1148 | |||
AK153970.1 | - | 10 | 1122 | |||
AK166687.1 | - | 4 | 367 | |||
AK168246.1 | - | 10 | 1154 | |||
BC019500.1 | - | 10 | 1122 | 3602929 | ||
D13545.1 | - | 10 | 1125 | |||
D17385.1 | - | 10 | 1105 |