Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkar2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011158.3 | NP_035288.2 | 11 | 1385 | ||
GenBank mRNA | AK041013.1 | - | 11 | 1359 | ||
AK081805.1 | - | 11 | 1162 | |||
AK135621.1 | - | 11 | 1385 | |||
AK138963.1 | - | 11 | 1351 | |||
BC033267.1 | - | 7 | 673 | |||
BC048710.1 | - | 11 | 1341 | 6506827 |