Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkcsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293650.1 | NP_001280579.1 | 5, 6 | 517 | ||
NM_001293651.1 | NP_001280580.1 | 5, 6 | 521 | |||
NM_008925.2 | NP_032951.1 | 5, 6 | 517 | |||
GenBank mRNA | AK137661.1 | - | 5, 6 | 484 | ||
AK153931.1 | - | 5, 6 | 492 | |||
AK159705.1 | - | 5, 6 | 488 | |||
BC009816.1 | - | 5, 6 | 450 | 4008182 | ||
CD807078.1 | - | 5, 6 | 486 | |||
U92794.1 | - | 5, 6 | 470 |