Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008926.4 | NP_032952.3 | 3 | 776 | ||
GenBank mRNA | AK038739.1 | - | 3 | 760 | ||
AK081422.1 | - | 3 | 777 | |||
BC113205.1 | - | 3 | 687 | 40047751 | ||
L12460.1 | - | 3 | 1048 |