Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008936.1 | NP_032962.1 | 1, 2 | 343 | ||
GenBank mRNA | U77946.1 | - | 1, 2 | 343 |