Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013830.2 | NP_038858.2 | 6 | 1996 | ||
GenBank mRNA | AB516304.1 | - | 6 | 1848 | ||
AF033663.1 | - | 5 | 1286 | |||
AF283466.1 | - | 6 | 1994 | |||
AK083926.1 | - | 6 | 1880 | |||
BC003769.1 | - | 3 | 373 | |||
BC042521.1 | - | 7 | 1278 | |||
BC080864.1 | - | 6 | 1967 | |||
BC141272.1 | - | 6 | 1922 | 9055884 | ||
BC141273.1 | - | 6 | 1922 | 9055885 | ||
U48737.1 | - | 4 | 553 |