Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgmn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011175.2 | NP_035305.1 | 4 | 357 | ||
GenBank mRNA | AF044266.1 | - | 4 | 356 | ||
AJ000990.1 | - | 4 | 357 | |||
AK149666.1 | - | 4 | 412 | |||
BC132515.1 | - | 3 | 304 | 40130792 | ||
BC132519.1 | - | 3 | 304 | 40130796 |