Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Helq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081107.1 | NP_001074576.1 | 2 | 541 | ||
GenBank mRNA | AK040335.1 | - | 2 | 525 | ||
AK050769.1 | - | 2 | 518 | |||
AK052427.1 | - | 2 | 501 | |||
BC082601.1 | - | 2 | 505 | 30662103 | ||
BC109169.2 | - | 2 | 409 | |||
BC109170.2 | - | 2 | 414 | 40048258 |