Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008947.3 | NP_032973.1 | 8, 9 | 1051 | ||
GenBank mRNA | AK088411.1 | - | 9, 10 | 1081 | ||
AK133926.1 | - | 9, 10 | 1085 | |||
AK148906.1 | - | 9, 10 | 1073 | |||
AK161641.1 | - | 9, 10 | 1100 | |||
AK161723.1 | - | 9, 10 | 1101 | |||
AK166783.1 | - | 9, 10 | 1066 | |||
AK169015.1 | - | 9, 10 | 1064 | |||
BC003860.1 | - | 9, 10 | 1067 | 3486436 | ||
U39302.1 | - | 8, 9 | 1052 |