Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138672.2 | NP_619613.2 | 56 | 6137 | ||
GenBank mRNA | AB052959.1 | - | 56 | 6058 | ||
AF290914.2 | - | 56 | 6068 | |||
BC019712.1 | - | 2 | 256 | |||
BC031166.1 | - | 19 | 1991 | |||
BC049247.1 | - | 7 | 1143 |