Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138677.2 | NP_619618.1 | 12 | 1997 | ||
GenBank mRNA | AB042828.1 | - | 12 | 1986 | ||
AK030943.1 | - | 12 | 2217 | |||
AK037889.1 | - | 12 | 1560 | |||
AK082920.1 | - | 12 | 1950 | |||
AK129090.1 | - | 12 | 1964 | |||
AK135969.1 | - | 7 | 861 | |||
AK151668.1 | - | 12 | 1972 | |||
AK152724.1 | - | 12 | 1973 | |||
AK152979.1 | - | 12 | 1969 | |||
AK193028.1 | - | 2 | 101 | |||
BC023237.1 | - | 12 | 1945 | 2654113 |