Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Luc7l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170848.1 | NP_001164319.1 | 6, 7 | 1083 | ||
NM_001170849.1 | NP_001164320.1 | 6, 7 | 1083 | |||
NM_138680.2 | NP_619621.2 | 6, 7 | 1083 | |||
GenBank mRNA | AF318301.1 | - | 6, 7 | 1008 | ||
AK046307.1 | - | 7, 8 | 1281 | |||
AK134191.1 | - | 6, 7 | 997 | |||
AK147889.1 | - | 9, 10 | 1254 | |||
BC002314.1 | - | 1, 2 | 18 | |||
BC020058.1 | - | 7, 8 | 808 | 4009362 | ||
BC040793.1 | - | 5, 6 | 575 | |||
BC054449.1 | - | 5, 6 | 1026 | |||
BC056354.1 | - | 6, 7 | 1081 | 6811460 | ||
BC056383.1 | - | 6, 7 | 1062 | 6853955 | ||
BC056970.1 | - | 6, 7 | 1081 | 6811460 |