Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf21a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109690.1 | NP_001103160.1 | 1 | 19 | ||
NM_001109691.1 | NP_001103161.1 | 5 | 427 | |||
NM_138755.2 | NP_620094.2 | 4 | 433 | |||
GenBank mRNA | AB105178.1 | - | 4 | 339 | ||
AK037174.1 | - | 5 | 428 | |||
AK129424.1 | - | 3 | 1437 | |||
BC019181.1 | - | 4 | 288 | 5008954 | ||
BY303333.1 | - | 3 | 202 | |||
CX217243.1 | - | 4 | 433 |