Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162365.1 | NP_001155837.1 | 21 | 2112 | ||
NM_001162366.1 | NP_001155838.1 | 20 | 1942 | |||
NM_172498.3 | NP_766086.2 | 21 | 2112 | |||
GenBank mRNA | AK088696.1 | - | 21 | 2113 | ||
AK150105.1 | - | 21 | 2043 | |||
BC024594.1 | - | 2 | 116 | |||
BC137704.1 | - | 20 | 1937 | 8860723 | ||
BC144849.1 | - | 20 | 1937 | 9053397 | ||
JN971017.1 | - | 20 | 1905 |