Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166388.1 | NP_001159860.1 | 6 | 1941 | ||
NM_001166389.1 | NP_001159861.1 | 6 | 1672 | |||
NM_001166390.1 | NP_001159862.1 | 5 | 773 | |||
NM_008975.3 | NP_033001.2 | 6 | 1741 | |||
GenBank mRNA | AF035645.1 | - | 5 | 722 | ||
AK003954.1 | - | 5 | 479 | |||
AK004562.1 | - | 6 | 1350 | |||
AK014601.1 | - | 6 | 1355 | |||
AK143702.1 | - | 6 | 1393 | |||
AK147489.1 | - | 4 | 6134 | |||
AK172192.1 | - | 6 | 1395 | |||
AK190358.1 | - | 1 | 51 | |||
BC027445.1 | - | 5 | 552 | 4482106 | ||
BC066043.1 | - | 6 | 1941 | 6415021 | ||
CT010215.1 | - | 5 | 507 |