Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008976.2 | NP_033002.2 | 7 | 1138 | ||
GenBank mRNA | AF170902.1 | - | 6 | 705 | ||
AF170903.1 | - | 6 | 705 | |||
AK028783.1 | - | 7 | 1102 | |||
AK037646.1 | - | 7 | 1139 | |||
AK086524.1 | - | 7 | 1132 | |||
AK156480.1 | - | 7 | 884 | |||
D31842.1 | - | 6 | 735 |
Gene Symbol: Gm26574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK143393.1 | - | 2 | 1941 |