Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011206.2 | NP_035336.2 | 8 | 684 | ||
GenBank mRNA | AK144353.1 | - | 8 | 666 | ||
BC008512.1 | - | 8 | 654 | 3591418 | ||
U35124.1 | - | 8 | 658 | |||
U49853.1 | - | 8 | 624 | |||
U52523.1 | - | 8 | 672 |