Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfpl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145013.1 | NP_001138485.1 | 3 | 532 | ||
NM_138954.3 | NP_620404.1 | 3 | 764 | |||
GenBank mRNA | AK136022.1 | - | 3 | 533 | ||
AK139569.1 | - | 3 | 767 | |||
AY070253.1 | - | 3 | 754 | |||
BC069834.1 | - | 3 | 518 | 30474534 |