Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138955.3 | NP_620405.3 | 7 | 1037 | ||
GenBank mRNA | AF378330.1 | - | 6 | 868 | ||
AF411084.1 | - | 7 | 915 | |||
AF425077.1 | - | 7 | 917 | |||
AJ426047.2 | - | 7 | 916 | |||
AK043147.1 | - | 7 | 1038 | |||
AK138415.1 | - | 6 | 878 | |||
AY040865.1 | - | 7 | 881 | |||
BC016200.1 | - | 7 | 917 | 4511456 | ||
BC023077.1 | - | 1 | 125 | |||
BC026477.1 | - | 252 |