Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138955.3 | NP_620405.3 | 15 | 2169 | ||
GenBank mRNA | AF378330.1 | - | 14 | 2000 | ||
AF411084.1 | - | 15 | 2047 | |||
AF425077.1 | - | 15 | 2049 | |||
AJ426047.2 | - | 15 | 2048 | |||
AK043147.1 | - | 15 | 2170 | |||
AK157317.1 | - | 15 | 2494 | |||
AY040865.1 | - | 15 | 2013 | |||
BC016200.1 | - | 15 | 2049 | 4511456 | ||
BC023077.1 | - | 9 | 1257 | |||
BC030301.1 | - | 1 | 365 | |||
BC059033.1 | - | 6 | 1342 |