Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008983.2 | NP_033009.1 | 6 | 1058 | ||
GenBank mRNA | AK019870.1 | - | 6 | 958 | ||
AK052275.1 | - | 6 | 1012 | |||
AK083006.1 | - | 6 | 1022 | |||
BC138376.1 | - | 6 | 1024 | 8861396 | ||
BC145261.1 | - | 6 | 1024 |