Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpru | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083119.2 | NP_001076588.1 | 8 | 1353 | ||
NM_011214.2 | NP_035344.2 | 8 | 1353 | |||
GenBank mRNA | AK047640.1 | - | 7 | 1051 | ||
AK052720.1 | - | 8 | 1328 | |||
AK148515.1 | - | 8 | 1350 | |||
BC080736.1 | - | 8 | 1308 | |||
D88187.1 | - | 8 | 1344 |