Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161837.1 | NP_001155309.1 | 11 | 1436 | ||
NM_001161838.1 | NP_001155310.1 | 8 | 1393 | |||
NM_001161839.1 | NP_001155311.1 | 9 | 1084 | |||
NM_001161840.1 | NP_001155312.1 | 9 | 1276 | |||
NM_011217.2 | NP_035347.1 | 12 | 2042 | |||
GenBank mRNA | AF041866.1 | - | 9 | 1201 | ||
AF129509.1 | - | 8 | 1318 | |||
AK028054.1 | - | 12 | 2043 | |||
BC119230.1 | - | 12 | 1975 | 8733979 | ||
BC120547.1 | - | 12 | 1975 | 40129470 | ||
D31898.1 | - | 12 | 2005 | |||
Z30313.1 | - | 11 | 1431 |