Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161837.1 | NP_001155309.1 | 12 | 1570 | ||
NM_001161838.1 | NP_001155310.1 | 9 | 1527 | |||
NM_001161839.1 | NP_001155311.1 | 10 | 1218 | |||
NM_001161840.1 | NP_001155312.1 | 10 | 1410 | |||
NM_011217.2 | NP_035347.1 | 13 | 2176 | |||
GenBank mRNA | AF041866.1 | - | 10 | 1335 | ||
AF129509.1 | - | 9 | 1452 | |||
AK028054.1 | - | 13 | 2177 | |||
BC119230.1 | - | 13 | 2109 | 8733979 | ||
BC120547.1 | - | 13 | 2109 | 40129470 | ||
D31898.1 | - | 13 | 2139 | |||
Z30313.1 | - | 12 | 1565 |