Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005608.2 | NP_001005608.2 | 8 | 1286 | ||
NM_133663.2 | NP_598424.2 | 8 | 1286 | |||
GenBank mRNA | AK048888.1 | - | 8 | 1168 | ||
AK142648.1 | - | 8 | 976 | |||
AK147424.1 | - | 8 | 1286 | |||
BC059192.1 | - | 4 | 478 | |||
L04678.1 | - | 9 | 1133 |