Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvrl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159724.1 | NP_001153196.1 | 2 | 675 | ||
NM_008990.3 | NP_033016.3 | 2 | 675 | |||
GenBank mRNA | AK075815.1 | - | 2 | 580 | ||
AK157513.1 | - | 2 | 580 | |||
BC009088.1 | - | 2 | 568 | 2812478 | ||
BC046770.1 | - | 2 | 571 | 5703547 | ||
BC059941.1 | - | 2 | 627 | 6530953 | ||
D26107.1 | - | 2 | 404 | |||
M80206.1 | - | 2 | 404 |