Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008992.2 | NP_033018.2 | 13, 14 | 1559 | ||
GenBank mRNA | AF213384.1 | - | 761 | |||
AJ001166.1 | - | 13, 14 | 1463 | |||
AK028420.1 | - | 13, 14 | 1616 | |||
BC050102.1 | - | 13, 14 | 1667 | 30012556 |