Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278447.1 | NP_001265376.1 | 6 | 532 | ||
NM_181070.6 | NP_851415.1 | 6 | 514 | |||
GenBank mRNA | AB232612.1 | - | 6 | 416 | ||
AK028155.1 | - | 2 | 115 | |||
AK045514.1 | - | 6 | 477 | |||
AK140901.1 | - | 6 | 482 | |||
AK143673.1 | - | 6 | 490 | |||
AK146177.1 | - | 6 | 439 | |||
AK146397.1 | - | 6 | 439 | |||
AK154920.1 | - | 6 | 491 | |||
BC056351.1 | - | 6 | 468 | 6807284 | ||
L04966.1 | - | 6 | 420 | |||
X80333.1 | - | 6 | 416 |