Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024436.3 | NP_077756.2 | 2 | 361 | ||
GenBank mRNA | AB232616.1 | - | 2 | 100 | ||
AJ311124.1 | - | 2 | 157 | |||
AK014407.1 | - | 347 | ||||
AK031667.1 | - | 2 | 341 | |||
AK146594.1 | - | 2 | 361 | |||
AK147619.1 | - | 447 | ||||
AK149200.1 | - | 353 | ||||
AK150617.1 | - | 2 | 367 | |||
AK219950.1 | - | 2 | 106 | |||
BC006596.1 | - | 2 | 348 | 3965264 | ||
M79304.1 | - | 1 | 52 |