Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab33b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016858.2 | NP_058554.1 | 1 | 468 | ||
GenBank mRNA | AB004665.1 | - | 1 | 418 | ||
AB232629.1 | - | 1 | 102 | |||
AK004974.1 | - | 1 | 204 | |||
AK087967.1 | - | 1 | 208 | |||
AK139821.1 | - | 1 | 454 | |||
AK143171.1 | - | 394 | ||||
AK159674.1 | - | 1 | 203 | |||
AK163783.1 | - | 1 | 455 | |||
AK170483.1 | - | 1 | 97 | |||
BC065076.1 | - | 1 | 468 | 6815992 |