Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044371.2 | NP_001037836.1 | 4 | 536 | ||
NM_001283011.1 | NP_001269940.1 | 4 | 536 | |||
NM_011233.3 | NP_035363.2 | 4 | 664 | |||
GenBank mRNA | AF085737.1 | - | 4 | 587 | ||
AJ011923.1 | - | 3 | 437 | |||
AK134277.1 | - | 4 | 491 | |||
AK162016.1 | - | 4 | 523 | |||
AK167358.1 | - | 4 | 479 | |||
BC023939.1 | - | 4 | 541 | |||
BC050267.1 | - | 1 | 176 | |||
BC066855.1 | - | 4 | 654 | 30473984 |