Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159482.1 | NP_001152954.1 | 9, 10 | 853 | ||
NM_033475.3 | NP_258436.2 | 8, 9 | 1198 | |||
GenBank mRNA | AB082927.1 | - | 8, 9 | 1111 | ||
AB232630.1 | - | 8, 9 | 609 | |||
AF322068.1 | - | 8, 9 | 609 | |||
AF327929.1 | - | 9, 10 | 803 | |||
AK160308.1 | - | 1167 | ||||
AK181473.1 | - | 1, 2 | 23 | |||
AK188927.1 | - | 1, 2 | 23 | |||
BC038638.1 | - | 8, 9 | 673 | 5058535 | ||
S72304.1 | - | 6, 7 | 456 |