Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159482.1 | NP_001152954.1 | 5 | 472 | ||
NM_033475.3 | NP_258436.2 | 4 | 817 | |||
GenBank mRNA | AB082927.1 | - | 4 | 730 | ||
AB232630.1 | - | 4 | 228 | |||
AF322068.1 | - | 4 | 228 | |||
AF327929.1 | - | 5 | 422 | |||
AK160308.1 | - | 786 | ||||
AK179020.1 | - | 2 | 100 | |||
AK200345.1 | - | 3 | 148 | |||
AK205613.1 | - | 3 | 99 | |||
AK205647.1 | - | 3 | 99 | |||
AK216992.1 | - | 3 | 99 | |||
BC038638.1 | - | 4 | 292 | 5058535 | ||
S72304.1 | - | 2 | 75 |