Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rangap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146174.1 | NP_001139646.1 | 5 | 678 | ||
NM_011241.4 | NP_035371.4 | 6 | 726 | |||
GenBank mRNA | AK035571.1 | - | 6 | 688 | ||
AK042930.1 | - | 5 | 643 | |||
AK088768.1 | - | 5 | 600 | |||
AK129452.1 | - | 5 | 607 | |||
AK133905.1 | - | 5 | 644 | |||
AK146670.1 | - | 6 | 686 | |||
AK159009.1 | - | 5 | 631 | |||
AK159525.1 | - | 5 | 643 | |||
AK159930.1 | - | 5 | 634 | |||
AK178591.1 | - | 4 | 357 | |||
AK178655.1 | - | 4 | 358 | |||
AK181740.1 | - | 4 | 358 | |||
AK205227.1 | - | 3 | 308 | |||
BC014855.1 | - | 5 | 634 | 3157225 | ||
BC052862.1 | - | 5 | 601 | 30063509 | ||
BC066213.1 | - | 5 | 642 | 30466595 | ||
BC071200.1 | - | 6 | 842 | 5032610 | ||
U08110.1 | - | 6 | 709 |