Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rangap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146174.1 | NP_001139646.1 | 8, 9 | 1126 | ||
NM_011241.4 | NP_035371.4 | 9, 10 | 1174 | |||
GenBank mRNA | AK042930.1 | - | 8, 9 | 1092 | ||
AK088768.1 | - | 8, 9 | 1048 | |||
AK129452.1 | - | 8, 9 | 1055 | |||
AK133905.1 | - | 8, 9 | 1094 | |||
AK145459.1 | - | 7, 8 | 729 | |||
AK146670.1 | - | 9, 10 | 1134 | |||
AK159009.1 | - | 8, 9 | 1079 | |||
AK159525.1 | - | 8, 9 | 1091 | |||
AK159930.1 | - | 8, 9 | 1082 | |||
AK205795.1 | - | 1, 2 | 86 | |||
AK213698.1 | - | 1, 2 | 74 | |||
BC014855.1 | - | 8, 9 | 1082 | 3157225 | ||
BC052862.1 | - | 8, 9 | 1049 | 30063509 | ||
BC066213.1 | - | 8, 9 | 1090 | 30466595 | ||
BC071200.1 | - | 9, 10 | 1290 | 5032610 | ||
U08110.1 | - | 9, 10 | 1157 |