Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rarg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042727.2 | NP_001036192.1 | 8 | 1509 | ||
NM_011244.4 | NP_035374.3 | 10 | 1747 | |||
GenBank mRNA | AK148064.1 | - | 10 | 1744 | ||
AK155911.1 | - | 8 | 1363 | |||
BC012923.1 | - | 10 | 1673 | 3154037 | ||
BC013709.1 | - | 8 | 1301 | 3989967 | ||
M34475.1 | - | 8 | 1285 | |||
M34476.1 | - | 10 | 1504 | |||
X15848.1 | - | 10 | 1567 |