Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgrf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009027.3 | NP_033053.2 | 3161 | |||
GenBank mRNA | AK158472.1 | - | 3326 | |||
U67326.2 | - | 3161 |