Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnksr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081047.1 | NP_001074516.1 | 21 | 2011 | ||
GenBank mRNA | AK145910.1 | - | 21 | 1973 | ||
BC037758.1 | - | 2 | 241 | |||
BC113805.1 | - | 21 | 1976 | 40062208 | ||
BC113956.1 | - | 21 | 1976 | 40062209 |